siRNA Details
| Gene Symbol: Dnajc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025384.3 | NP_079660.1 | 4 | 308 | ||
| GenBank mRNA | AK002598.1 | - | 4 | 304 | ||
| AK003396.1 | - | 4 | 318 | |||
| AK007806.1 | - | 4 | 297 | |||
| AK007833.1 | - | 4 | 297 | |||
| BC027509.1 | - | 4 | 308 | 1364626 | ||

Dnajc15
Genomic Map