siRNA Details
| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164472.1 | NP_001157944.1 | 4 | 1077 | ||
| NM_025391.2 | NP_079667.2 | 5 | 1218 | |||
| GenBank mRNA | AK003739.1 | - | 5 | 1218 | ||
| AK014290.1 | - | 5 | 1177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nip7| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164472.1 | NP_001157944.1 | 4 | 1077 | ||
| NM_025391.2 | NP_079667.2 | 5 | 1218 | |||
| GenBank mRNA | AK003739.1 | - | 5 | 1218 | ||
| AK014290.1 | - | 5 | 1177 | |||