siRNA Details
| Gene Symbol: Alg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025442.3 | NP_079718.1 | 2 | 176 | ||
| GenBank mRNA | AK003818.1 | - | 2 | 157 | ||
| AK005296.1 | - | 2 | 177 | |||
| AK011159.1 | - | 2 | 145 | |||
| AK030567.1 | - | 2 | 149 | |||
| AK161187.1 | - | 157 | ||||
| BC027160.1 | - | 2 | 136 | 4946430 | ||

Alg5
Genomic Map