siRNA Details
| Gene Symbol: Mptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205011.2 | NP_001191940.1 | 2 | 114 | ||
| Gene Symbol: Mptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025470.3 | NP_079746.1 | 2 | 135 | ||
| GenBank mRNA | AK007658.1 | - | 2 | 136 | ||
| AK007963.1 | - | 2 | 135 | |||

Mptx2
Genomic Map