siRNA Details
| Gene Symbol: Tmem128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025480.3 | NP_079756.2 | 2 | 152 | ||
| GenBank mRNA | AK012789.2 | - | 2 | 153 | ||
| AK013227.1 | - | 2 | 152 | |||
| AK076679.1 | - | 2 | 144 | |||
| AK080853.1 | - | 2 | 153 | |||
| AK162257.1 | - | 2 | 147 | |||
| BC025197.1 | - | 2 | 311 | 5251991 | ||
| BC060969.1 | - | 2 | 141 | 6704414 | ||
| BC110310.1 | - | 2 | 135 | 5325405 | ||

Tmem128
Genomic Map