siRNA Details
| Gene Symbol: Dnajc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163536.1 | NP_001157008.1 | 2 | 205 | ||
| NM_001163537.1 | NP_001157009.1 | 2 | 225 | |||
| NM_025489.3 | NP_079765.3 | 2 | 212 | |||
| GenBank mRNA | AA426920.1 | - | 2 | 279 | ||
| AK005725.1 | - | 2 | 192 | |||
| AK005749.1 | - | 2 | 181 | |||
| BC049579.1 | - | 2 | 155 | 6772915 | ||
| BY085815.1 | - | 2 | 206 | |||

Dnajc5b
Genomic Map