siRNA Details
| Gene Symbol: Cenpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025495.3 | NP_079771.1 | 2 | 194 | ||
| GenBank mRNA | AK006234.1 | - | 2 | 231 | ||
| AK012863.1 | - | 2 | 198 | |||
| AK014919.1 | - | 2 | 150 | |||
| AK076577.1 | - | 2 | 232 | |||
| BC094280.1 | - | 2 | 248 | |||
| BC147207.1 | - | 2 | 174 | 9056262 | ||
| BC147208.1 | - | 2 | 174 | 9056260 | ||

Cenpp
Genomic Map