siRNA Details
| Gene Symbol: Ergic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025516.4 | NP_079792.1 | 6 | 600 | ||
| GenBank mRNA | AK009350.1 | - | 6 | 601 | ||
| AK013942.1 | - | 6 | 600 | |||
| AK050285.1 | - | 6 | 615 | |||
| AK190333.1 | - | 2 | 176 | |||
| BC043720.1 | - | 6 | 625 | 5375947 | ||
| BC057130.1 | - | 6 | 619 | 6819420 | ||

Ergic3
Genomic Map