siRNA Details
| Gene Symbol: Rnf113a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025525.2 | NP_079801.2 | 1 | 537 | ||
| GenBank mRNA | AK009420.1 | - | 1 | 557 | ||
| AK016795.1 | - | 1 | 497 | |||
| BC058194.1 | - | 1 | 443 | |||
| BC116426.2 | - | 1 | 478 | 40104196 | ||

Rnf113a2
Genomic Map