siRNA Details
| Gene Symbol: 1110059G10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025419.4 | NP_079695.2 | 2 | 309 | ||
| GenBank mRNA | AK004312.1 | - | 2 | 305 | ||
| AK012115.1 | - | 2 | 305 | |||
| AK083316.1 | - | 2 | 282 | |||
| AK160617.1 | - | 2 | 310 | |||
| BC034133.1 | - | 2 | 267 | 4921846 | ||

1110059G10Rik
Genomic Map