siRNA Details
Gene Symbol: 1110059E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025423.2 | NP_079699.1 | 4 | 458 | ||
GenBank mRNA | AK004308.1 | - | 4 | 459 | ||
AK016266.1 | - | 4 | 605 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1110059E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025423.2 | NP_079699.1 | 4 | 458 | ||
GenBank mRNA | AK004308.1 | - | 4 | 459 | ||
AK016266.1 | - | 4 | 605 |