siRNA Details
| Gene Symbol: Mrps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025440.2 | NP_079716.1 | 3 | 512 | ||
| GenBank mRNA | AK005206.1 | - | 3 | 513 | ||
| AK019243.1 | - | 3 | 458 | |||
| BC024656.1 | - | 3 | 478 | 3986314 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps16| Gene Symbol: Mrps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025440.2 | NP_079716.1 | 3 | 512 | ||
| GenBank mRNA | AK005206.1 | - | 3 | 513 | ||
| AK019243.1 | - | 3 | 458 | |||
| BC024656.1 | - | 3 | 478 | 3986314 | ||