siRNA Details
| Gene Symbol: Ccdc28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025455.2 | NP_079731.2 | 3 | 407 | ||
| GenBank mRNA | AK003287.1 | - | 3 | 377 | ||
| AK007427.1 | - | 2 | 184 | |||
| AK009176.1 | - | 1 | 26 | |||
| AK028262.1 | - | 3 | 385 | |||
| AK031278.1 | - | 1 | 133 | |||
| AK158016.1 | - | 3 | 411 | |||
| BC028873.1 | - | 3 | 399 | 4220355 | ||

Ccdc28b
Genomic Map