siRNA Details
| Gene Symbol: Eapp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025456.3 | NP_079732.1 | 2 | 320 | ||
| GenBank mRNA | AK007442.1 | - | 2 | 275 | ||
| AK012800.1 | - | 2 | 277 | |||
| AY882557.1 | - | 2 | 239 | |||
| BC037622.1 | - | 2 | 347 | 5373275 | ||

Eapp
Genomic Map