siRNA Details
| Gene Symbol: Tmem126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025460.2 | NP_079736.1 | 3, 4 | 435 | ||
| GenBank mRNA | AK005343.1 | - | 3, 4 | 414 | ||
| AK007559.1 | - | 3, 4 | 436 | |||
| AK210038.1 | - | 1, 2 | 112 | |||
| BC023171.1 | - | 3, 4 | 399 | 4946998 | ||

Tmem126a
Genomic Map