siRNA Details
| Gene Symbol: Isoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025478.3 | NP_079754.2 | 3 | 451 | ||
| GenBank mRNA | AK011674.1 | - | 3 | 318 | ||
| AK134794.1 | - | 3 | 451 | |||
| AK165542.1 | - | 3 | 470 | |||
| AK185662.1 | - | 2 | 149 | |||
| AY035213.1 | - | 3 | 451 | |||
| BC016576.1 | - | 3 | 458 | 2609919 | ||

Isoc1
Genomic Map