siRNA Details
| Gene Symbol: Tmem128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025480.3 | NP_079756.2 | 3, 4 | 420 | ||
| GenBank mRNA | AK012789.2 | - | 3, 4 | 421 | ||
| AK013227.1 | - | 3, 4 | 420 | |||
| AK076679.1 | - | 3, 4 | 412 | |||
| AK080853.1 | - | 3, 4 | 421 | |||
| AK162257.1 | - | 3, 4 | 415 | |||
| BC025197.1 | - | 3, 4 | 579 | 5251991 | ||
| BC060969.1 | - | 3, 4 | 409 | 6704414 | ||
| BC110310.1 | - | 3, 4 | 403 | 5325405 | ||

Tmem128
Genomic Map