siRNA Details
| Gene Symbol: Cenpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025495.3 | NP_079771.1 | 3, 4 | 410 | ||
| GenBank mRNA | AK006234.1 | - | 3, 4 | 447 | ||
| AK012863.1 | - | 3, 4 | 414 | |||
| AK014919.1 | - | 3, 4 | 366 | |||
| AK076577.1 | - | 3, 4 | 448 | |||
| BC094280.1 | - | 3, 4 | 463 | |||
| BC147207.1 | - | 3, 4 | 390 | 9056262 | ||
| BC147208.1 | - | 3, 4 | 390 | 9056260 | ||

Cenpp
Genomic Map