siRNA Details
| Gene Symbol: Ergic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025516.4 | NP_079792.1 | 6, 7 | 651 | ||
| GenBank mRNA | AK009350.1 | - | 6, 7 | 652 | ||
| AK050285.1 | - | 6, 7 | 666 | |||
| AK190333.1 | - | 2, 3 | 227 | |||
| BC043720.1 | - | 6, 7 | 676 | 5375947 | ||
| BC057130.1 | - | 6, 7 | 670 | 6819420 | ||

Ergic3
Genomic Map