siRNA Details
| Gene Symbol: Rtca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025517.3 | NP_079793.2 | 4 | 506 | ||
| GenBank mRNA | AK002543.1 | - | 4 | 417 | ||
| AK009245.1 | - | 4 | 442 | |||
| AK151108.1 | - | 4 | 394 | |||
| AK151400.1 | - | 4 | 394 | |||
| AK152129.1 | - | 4 | 507 | |||
| AK159961.1 | - | 4 | 396 | |||
| AK167830.1 | - | 4 | 383 | |||
| AK179135.1 | - | 2 | 129 | |||
| BC016519.1 | - | 4 | 432 | 3495638 | ||
| BC090403.1 | - | 4 | 477 | 5293337 | ||

Rtca
Genomic Map