siRNA Details
| Gene Symbol: Zdhhc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037762.1 | NP_001032851.1 | 5 | 1009 | ||
| NM_025428.2 | NP_079704.2 | 5 | 1051 | |||
| GenBank mRNA | AK004475.1 | - | 5 | 952 | ||
| AK013074.1 | - | 5 | 1038 | |||
| AK087676.1 | - | 5 | 1023 | |||
| BC021432.1 | - | 5 | 991 | 5030263 | ||

Zdhhc12
Genomic Map