siRNA Details
| Gene Symbol: Alg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025442.3 | NP_079718.1 | 10 | 1311 | ||
| GenBank mRNA | AK005296.1 | - | 10 | 1312 | ||
| AK161187.1 | - | 1289 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg5| Gene Symbol: Alg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025442.3 | NP_079718.1 | 10 | 1311 | ||
| GenBank mRNA | AK005296.1 | - | 10 | 1312 | ||
| AK161187.1 | - | 1289 | ||||