siRNA Details
| Gene Symbol: Ing5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025454.2 | NP_079730.1 | 7, 8 | 719 | ||
| GenBank mRNA | AK006421.1 | - | 7, 8 | 818 | ||
| AK007536.1 | - | 7, 8 | 710 | |||
| AK028010.1 | - | 7, 8 | 697 | |||
| AK028397.1 | - | 7, 8 | 720 | |||
| AK145695.1 | - | 7, 8 | 692 | |||
| BC064674.1 | - | 7, 8 | 718 | 6400183 | ||

Ing5
Genomic Map