siRNA Details
| Gene Symbol: Tmem126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025460.2 | NP_079736.1 | 5 | 704 | ||
| GenBank mRNA | AK007559.1 | - | 5 | 705 | ||
| AK050267.1 | - | 5 | 675 | |||
| BC023171.1 | - | 5 | 668 | 4946998 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem126a| Gene Symbol: Tmem126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025460.2 | NP_079736.1 | 5 | 704 | ||
| GenBank mRNA | AK007559.1 | - | 5 | 705 | ||
| AK050267.1 | - | 5 | 675 | |||
| BC023171.1 | - | 5 | 668 | 4946998 | ||