siRNA Details
| Gene Symbol: Mptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025470.3 | NP_079746.1 | 2 | 291 | ||
| GenBank mRNA | AK007658.1 | - | 2 | 292 | ||
| AK007963.1 | - | 2 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mptx1| Gene Symbol: Mptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025470.3 | NP_079746.1 | 2 | 291 | ||
| GenBank mRNA | AK007658.1 | - | 2 | 292 | ||
| AK007963.1 | - | 2 | 291 | |||