siRNA Details
| Gene Symbol: Smim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025471.2 | NP_079747.1 | 4 | 593 | ||
| GenBank mRNA | AK005941.1 | - | 4 | 596 | ||
| AK007303.1 | - | 3 | 351 | |||
| AK007660.1 | - | 3 | 371 | |||
| AK021236.1 | - | 3 | 388 | |||
| AK079613.1 | - | 3 | 382 | |||
| BC027508.1 | - | 3 | 341 | 1494969 | ||

Smim8
Genomic Map