siRNA Details
| Gene Symbol: Tmem208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025486.2 | NP_079762.1 | 6 | 628 | ||
| GenBank mRNA | AK002499.1 | - | 6 | 616 | ||
| AK005662.1 | - | 6 | 612 | |||
| AK218712.1 | - | 1 | 104 | |||
| BC005665.1 | - | 6 | 584 | |||
| BC024608.1 | - | 6 | 579 | 4242513 | ||

Tmem208
Genomic Map