siRNA Details
| Gene Symbol: Ergic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025516.4 | NP_079792.1 | 9 | 783 | ||
| GenBank mRNA | AK009350.1 | - | 9 | 784 | ||
| AK013942.1 | - | 9 | 783 | |||
| AK050285.1 | - | 9 | 798 | |||
| AK183374.1 | - | 4 | 240 | |||
| AK190333.1 | - | 5 | 359 | |||
| BC043720.1 | - | 9 | 808 | 5375947 | ||
| BC057130.1 | - | 10 | 835 | 6819420 | ||

Ergic3
Genomic Map