siRNA Details
| Gene Symbol: Rtca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025517.3 | NP_079793.2 | 6 | 753 | ||
| GenBank mRNA | AK002543.1 | - | 6 | 663 | ||
| AK009245.1 | - | 6 | 689 | |||
| AK151108.1 | - | 6 | 641 | |||
| AK151400.1 | - | 6 | 641 | |||
| AK152129.1 | - | 6 | 754 | |||
| AK159961.1 | - | 6 | 643 | |||
| AK167830.1 | - | 6 | 630 | |||
| AK215360.1 | - | 3 | 176 | |||
| BC016519.1 | - | 6 | 679 | 3495638 | ||
| BC090403.1 | - | 6 | 724 | 5293337 | ||

Rtca
Genomic Map