siRNA Details
| Gene Symbol: Sln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025540.2 | NP_079816.1 | 2 | 231 | ||
| GenBank mRNA | AK008863.1 | - | 2 | 234 | ||
| AK008896.1 | - | 2 | 232 | |||
| AK009005.1 | - | 2 | 234 | |||
| AK009809.1 | - | 319 | ||||
| BC028496.1 | - | 2 | 212 | 1397989 | ||
| CA460968.1 | - | 2 | 269 | |||

Sln
Genomic Map