siRNA Details
| Gene Symbol: 2810428I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025577.2 | NP_079853.1 | 2 | 118 | ||
| GenBank mRNA | AK010635.1 | - | 2 | 115 | ||
| AK013181.1 | - | 2 | 119 | |||
| AK160511.1 | - | 2 | 62 | |||
| BC028495.1 | - | 2 | 124 | 3370477 | ||

2810428I15Rik
Genomic Map