siRNA Details
| Gene Symbol: Lamtor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025605.3 | NP_079881.2 | 3, 4 | 427 | ||
| GenBank mRNA | AK003509.1 | - | 3, 4 | 319 | ||
| AK009320.1 | - | 3, 4 | 316 | |||
| AK010247.1 | - | 3, 4 | 360 | |||
| AK013389.1 | - | 3, 4 | 337 | |||
| AK019947.1 | - | 3, 4 | 278 | |||
| AK144476.1 | - | 3, 4 | 326 | |||
| BC020142.1 | - | 3, 4 | 333 | 3709051 | ||
| BC092062.1 | - | 3, 4 | 319 | 4024506 | ||
| BC096412.1 | - | 3, 4 | 299 | 5255882 | ||
| BC126973.1 | - | 3, 4 | 334 | 40100355 | ||

Lamtor1
Genomic Map