siRNA Details
| Gene Symbol: Mien1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025559.2 | NP_079835.1 | 3 | 294 | ||
| GenBank mRNA | AK002959.1 | - | 3 | 277 | ||
| AK007795.1 | - | 3 | 277 | |||
| AK009922.1 | - | 3 | 272 | |||
| AK041314.1 | - | 3 | 276 | |||
| BC021589.1 | - | 3 | 278 | 5100542 | ||
| CA530220.1 | - | 2 | 120 | |||

Mien1
Genomic Map