siRNA Details
| Gene Symbol: 2010012O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025563.3 | NP_079839.1 | 2 | 224 | ||
| GenBank mRNA | AK007626.1 | - | 2 | 211 | ||
| AK008215.1 | - | 2 | 219 | |||
| AK046524.1 | - | 2 | 224 | |||
| BC027506.1 | - | 2 | 196 | 1514880 | ||

2010012O05Rik
Genomic Map