siRNA Details
| Gene Symbol: Spc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003408.1 | - | 3 | 282 | ||
| AK011893.1 | - | 4 | 400 | |||
| AK146727.1 | - | 4 | 910 | |||
| AK151554.1 | - | 4 | 441 | |||
| BC027121.1 | - | 4 | 408 | 5359960 | ||
| BC033605.1 | - | 4 | 657 | 5369405 | ||
| CX222514.1 | - | 4 | 467 | |||

Spc25
Genomic Map