siRNA Details
| Gene Symbol: Pyurf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025574.3 | NP_079850.1 | 2 | 554 | ||
| GenBank mRNA | AK003713.1 | - | 2 | 555 | ||
| AK008978.1 | - | 2 | 554 | |||
| AK011515.1 | - | 2 | 491 | |||
| AK029193.1 | - | 530 | ||||
| AK032381.1 | - | 2 | 547 | |||
| BC029232.1 | - | 2 | 518 | 4164517 | ||

Pyurf
Genomic Map