siRNA Details
| Gene Symbol: Ptpmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025576.2 | NP_079852.1 | 3 | 616 | ||
| GenBank mRNA | AK003210.1 | - | 3 | 320 | ||
| AK012674.1 | - | 3 | 606 | |||
| AK014691.1 | - | 3 | 610 | |||
| AK151972.1 | - | 3 | 617 | |||
| BC026750.1 | - | 3 | 595 | 5135841 | ||

Ptpmt1
Genomic Map