siRNA Details
| Gene Symbol: Fam213b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025582.3 | NP_079858.2 | 2 | 283 | ||
| GenBank mRNA | AK005188.1 | - | 2 | 284 | ||
| BC030453.1 | - | 2 | 277 | 5401948 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam213b| Gene Symbol: Fam213b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025582.3 | NP_079858.2 | 2 | 283 | ||
| GenBank mRNA | AK005188.1 | - | 2 | 284 | ||
| BC030453.1 | - | 2 | 277 | 5401948 | ||