siRNA Details
Gene Symbol: 1700029F12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080777.2 | NP_001074246.1 | 1, 2 | 186 | ||
NM_025585.3 | NP_079861.1 | 1, 2 | 186 | |||
GenBank mRNA | AK006036.1 | - | 1, 2 | 174 | ||
AK006488.1 | - | 1, 2 | 154 | |||
BC049576.1 | - | 1, 2 | 160 | 6772947 |
