siRNA Details
| Gene Symbol: Fam136a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025591.2 | NP_079867.1 | 3 | 355 | ||
| GenBank mRNA | AK005280.1 | - | 3 | 315 | ||
| AK008550.1 | - | 3 | 321 | |||
| AK141279.1 | - | 3 | 315 | |||
| BC013621.1 | - | 3 | 303 | |||
| BC024049.1 | - | 3 | 355 | 5100000 | ||
| BC103775.1 | - | 3 | 356 | 6485116 | ||
| CT010180.1 | - | 3 | 305 | |||

Fam136a
Genomic Map