siRNA Details
| Gene Symbol: Rwdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025614.3 | NP_079890.1 | 3 | 402 | ||
| GenBank mRNA | AF503942.1 | - | 3 | 294 | ||
| AK003262.2 | - | 3 | 356 | |||
| AK005278.1 | - | 3 | 314 | |||
| AK009510.1 | - | 3 | 271 | |||
| AK011277.1 | - | 3 | 402 | |||
| AK012504.1 | - | 3 | 373 | |||
| BC062136.1 | - | 3 | 284 | 6508909 | ||

Rwdd1
Genomic Map