siRNA Details
| Gene Symbol: Aggf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025630.3 | NP_079906.2 | 6 | 1264 | ||
| GenBank mRNA | AK009533.1 | - | 2 | 227 | ||
| AK132575.1 | - | 6 | 1265 | |||
| AY500995.1 | - | 6 | 1237 | |||
| BC052410.1 | - | 6 | 1241 | 5716440 | ||

Aggf1
Genomic Map