siRNA Details
| Gene Symbol: Asf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025541.3 | NP_079817.1 | 2 | 443 | ||
| GenBank mRNA | AK007804.1 | - | 2 | 299 | ||
| AK010210.1 | - | 2 | 453 | |||
| AK012376.1 | - | 2 | 434 | |||
| AK031513.1 | - | 2 | 409 | |||
| AK088618.1 | - | 2 | 386 | |||
| BC027628.1 | - | 2 | 235 | 1178866 | ||

Asf1a
Genomic Map