siRNA Details
| Gene Symbol: Pyurf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025574.3 | NP_079850.1 | 2 | 1433 | ||
| GenBank mRNA | AK032381.1 | - | 2 | 1427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pyurf| Gene Symbol: Pyurf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025574.3 | NP_079850.1 | 2 | 1433 | ||
| GenBank mRNA | AK032381.1 | - | 2 | 1427 | ||