siRNA Details
| Gene Symbol: Mrps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025578.4 | NP_079854.2 | 4 | 1452 | ||
| GenBank mRNA | AK013196.1 | - | 4 | 1410 | ||
| AK131114.1 | - | 4 | 1381 | |||
| BC022953.1 | - | 4 | 1439 | 5126320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps25| Gene Symbol: Mrps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025578.4 | NP_079854.2 | 4 | 1452 | ||
| GenBank mRNA | AK013196.1 | - | 4 | 1410 | ||
| AK131114.1 | - | 4 | 1381 | |||
| BC022953.1 | - | 4 | 1439 | 5126320 | ||