siRNA Details
| Gene Symbol: 2810004N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025615.2 | NP_079891.2 | 3 | 491 | ||
| GenBank mRNA | AK011944.1 | - | 3 | 441 | ||
| AK012675.1 | - | 3 | 453 | |||
| AK013989.1 | - | 3 | 479 | |||
| AK017273.1 | - | 3 | 451 | |||
| AK143919.1 | - | 457 | ||||
| AK166668.1 | - | 3 | 495 | |||
| BC023675.1 | - | 3 | 590 | 5354472 | ||

2810004N23Rik
Genomic Map