siRNA Details
| Gene Symbol: Teddm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009566.1 | - | 1 | 1128 | ||
| AK009777.1 | - | 1 | 1070 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Teddm3| Gene Symbol: Teddm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009566.1 | - | 1 | 1128 | ||
| AK009777.1 | - | 1 | 1070 | |||