siRNA Details
| Gene Symbol: Crls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024385.1 | NP_001019556.1 | 7 | 1095 | ||
| NM_025646.3 | NP_079922.2 | 7 | 782 | |||
| GenBank mRNA | AK016165.1 | - | 7 | 783 | ||
| AK018180.1 | - | 7 | 1095 | |||
| BC048702.1 | - | 7 | 1076 | 6506241 | ||
| BC086619.1 | - | 7 | 1052 | |||

Crls1
Genomic Map