siRNA Details
| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044637.1 | - | 7 | 1219 | ||
| AK084031.1 | - | 7 | 1203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Farsa| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044637.1 | - | 7 | 1219 | ||
| AK084031.1 | - | 7 | 1203 | |||