siRNA Details
| Gene Symbol: Lrrc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159609.1 | NP_001153081.1 | 4 | 669 | ||
| NM_001159610.1 | NP_001153082.1 | 4 | 720 | |||
| NM_001159612.1 | NP_001153084.1 | 4 | 736 | |||
| NM_025657.3 | NP_079933.2 | 4 | 619 | |||
| GenBank mRNA | AK003037.1 | - | 4 | 526 | ||
| AK003592.1 | - | 4 | 637 | |||
| AK012637.1 | - | 4 | 734 | |||
| AK015684.1 | - | 4 | 671 | |||
| AK019566.1 | - | 4 | 731 | |||
| AK033084.1 | - | 1 | 1536 | |||
| AK134971.1 | - | 4 | 578 | |||
| AK169249.1 | - | 4 | 643 | |||
| BC034894.1 | - | 4 | 578 | 3328402 | ||
| BU604191.1 | - | 4 | 622 | |||

Lrrc57
Genomic Map